siRNA Details
| Gene Symbol: Steap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107846.1 | NP_001101316.1 | 6 | 1685 | ||
| XM_006236032.2 | XP_006236094.1 | 6 | 1751 | |||
| XM_006236033.2 | XP_006236095.1 | 6 | 2213 | |||
| XM_006236034.2 | XP_006236096.1 | 6 | 1899 | |||
| XM_006236035.1 | XP_006236097.1 | 6 | 1640 | |||
| XM_006236036.2 | XP_006236098.1 | 6 | 1587 | |||
| XM_008762703.1 | XP_008760925.1 | 7 | 1942 | |||

Steap2
Genomic Map