siRNA Details
Gene Symbol: Golga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107847.1 | NP_001101317.1 | 2 | 639 | ||
XM_006249563.2 | XP_006249625.1 | 2 | 560 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Golga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107847.1 | NP_001101317.1 | 2 | 639 | ||
XM_006249563.2 | XP_006249625.1 | 2 | 560 |