siRNA Details
| Gene Symbol: Pon3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004086.1 | NP_001004086.1 | 7 | 743 | ||
| GenBank mRNA | BC079466.1 | - | 7 | 743 | 7122531 | |
| FQ218322.1 | - | 4 | 485 | |||
| FQ218352.1 | - | 4 | 482 | |||
| FQ218880.1 | - | 4 | 482 | |||
| FQ218895.1 | - | 4 | 483 | |||
| FQ219524.1 | - | 4 | 482 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173006, AK088572, L76193, AK076090, BC005714, and others.

Pon3
Genomic Map