siRNA Details
| Gene Symbol: Pon3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004086.1 | NP_001004086.1 | 1, 2 | 88 | ||
| GenBank mRNA | BC079466.1 | - | 1, 2 | 88 | 7122531 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pon3| Gene Symbol: Pon3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004086.1 | NP_001004086.1 | 1, 2 | 88 | ||
| GenBank mRNA | BC079466.1 | - | 1, 2 | 88 | 7122531 | |