siRNA Details
Gene Symbol: Vsig4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025004.1 | NP_001020175.1 | 3, 4 | 522 | ||
GenBank mRNA | BC091234.1 | - | 3, 4 | 522 | 7322659 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vsig4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025004.1 | NP_001020175.1 | 3, 4 | 522 | ||
GenBank mRNA | BC091234.1 | - | 3, 4 | 522 | 7322659 |