siRNA Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004267.1 | NP_001004267.1 | 6 | 671 | ||
| GenBank mRNA | AY387087.1 | - | 874 | |||
| BC079127.1 | - | 6 | 671 | 7127112 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem106b| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004267.1 | NP_001004267.1 | 6 | 671 | ||
| GenBank mRNA | AY387087.1 | - | 874 | |||
| BC079127.1 | - | 6 | 671 | 7127112 | ||