siRNA Details
| Gene Symbol: Ing3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034107.1 | NP_001029279.1 | 6 | 478 | ||
| XM_006236140.2 | XP_006236202.1 | 6 | 553 | |||
| XM_006236141.2 | XP_006236203.1 | 5 | 481 | |||
| GenBank mRNA | BC100082.1 | - | 6 | 478 | 7385392 | |

Ing3
Genomic Map