siRNA Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107850.1 | NP_001101320.1 | 5 | 608 | ||
| XM_006236190.2 | XP_006236252.1 | 6 | 784 | |||
| GenBank mRNA | FQ211470.1 | - | 5 | 549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm28| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107850.1 | NP_001101320.1 | 5 | 608 | ||
| XM_006236190.2 | XP_006236252.1 | 6 | 784 | |||
| GenBank mRNA | FQ211470.1 | - | 5 | 549 | ||