siRNA Details
| Gene Symbol: Tmem209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014055.1 | NP_001014077.1 | 8, 9 | 1062 | ||
| XM_006236202.2 | XP_006236264.1 | 8, 9 | 1329 | |||
| XM_006236204.2 | XP_006236266.1 | 8, 9 | 1133 | |||
| XM_006236205.1 | XP_006236267.1 | 8, 9 | 1180 | |||
| XM_006236207.2 | XP_006236269.2 | 8, 9 | 1191 | |||
| GenBank mRNA | BC079399.1 | - | 8, 9 | 1062 | 7112633 | |

Tmem209
Genomic Map