siRNA Details
| Gene Symbol: Plxna4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762859.1 | XP_008761081.1 | 23 | 3318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxna4a| Gene Symbol: Plxna4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762859.1 | XP_008761081.1 | 23 | 3318 | ||