siRNA Details
| Gene Symbol: Cnot4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037782.1 | NP_001032871.1 | 10 | 1503 | ||
| XM_006236271.2 | XP_006236333.1 | 10 | 1521 | |||
| XM_006236272.2 | XP_006236334.1 | 10 | 1530 | |||
| XM_006236273.2 | XP_006236335.1 | 10 | 1519 | |||
| XM_006236274.2 | XP_006236336.1 | 10 | 1528 | |||
| XM_008762777.1 | XP_008760999.1 | 10 | 1532 | |||
| GenBank mRNA | BC100152.1 | - | 10 | 1503 | 7456520 | |

Cnot4
Genomic Map