siRNA Details
| Gene Symbol: Adck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107855.1 | NP_001101325.1 | 1 | 950 | ||
| XM_006236325.2 | XP_006236387.1 | 1 | 974 | |||
| XM_006236326.2 | XP_006236388.1 | 1 | 973 | |||
| XM_008762782.1 | XP_008761004.1 | 1 | 973 | |||
| GenBank mRNA | BC161918.1 | - | 1 | 885 | 8369971 | |

Adck2
Genomic Map