siRNA Details
| Gene Symbol: Ppm1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107863.1 | NP_001101333.1 | 5 | 1115 | ||
| XM_006236571.2 | XP_006236633.1 | 6 | 1446 | |||
| GenBank mRNA | FQ211521.1 | - | 6 | 1238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppm1k| Gene Symbol: Ppm1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107863.1 | NP_001101333.1 | 5 | 1115 | ||
| XM_006236571.2 | XP_006236633.1 | 6 | 1446 | |||
| GenBank mRNA | FQ211521.1 | - | 6 | 1238 | ||