siRNA Details
| Gene Symbol: Immt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034928.1 | NP_001030100.1 | 5 | 579 | ||
| XM_008762981.1 | XP_008761203.1 | 5 | 713 | |||
| XM_008762982.1 | XP_008761204.1 | 5 | 713 | |||
| XM_008762983.1 | XP_008761205.1 | 5 | 679 | |||
| XM_008762984.1 | XP_008761206.1 | 5 | 676 | |||
| XM_008762985.1 | XP_008761207.1 | 5 | 679 | |||
| XM_008762986.1 | XP_008761208.1 | 5 | 711 | |||
| XM_008762987.1 | XP_008761209.1 | 5 | 677 | |||
| XM_008762988.1 | XP_008761210.1 | 5 | 674 | |||
| XM_008762989.1 | XP_008761211.1 | 5 | 677 | |||
| GenBank mRNA | BC083835.1 | - | 534 | |||
| BC105841.1 | - | 5 | 579 | 7454026 | ||

Immt
Genomic Map