siRNA Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025416.1 | NP_001020587.1 | 2, 3 | 360 | ||
| XM_008763022.1 | XP_008761244.1 | 2, 3 | 402 | |||
| GenBank mRNA | BC097972.1 | - | 2, 3 | 360 | 7442066 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dok1| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025416.1 | NP_001020587.1 | 2, 3 | 360 | ||
| XM_008763022.1 | XP_008761244.1 | 2, 3 | 402 | |||
| GenBank mRNA | BC097972.1 | - | 2, 3 | 360 | 7442066 | |