siRNA Details
| Gene Symbol: Rbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107875.1 | NP_001101345.1 | 3 | 507 | ||
| XM_006236908.2 | XP_006236970.1 | 3 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbsn| Gene Symbol: Rbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107875.1 | NP_001101345.1 | 3 | 507 | ||
| XM_006236908.2 | XP_006236970.1 | 3 | 522 | |||