siRNA Details
Gene Symbol: Pdzrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271251.1 | NP_001258180.1 | 4 | 1030 | ||
GenBank mRNA | FM040449.1 | - | 2 | 229 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdzrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271251.1 | NP_001258180.1 | 4 | 1030 | ||
GenBank mRNA | FM040449.1 | - | 2 | 229 |