siRNA Details
| Gene Symbol: Atg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012097.1 | NP_001012097.1 | 10 | 1006 | ||
| XM_006237033.2 | XP_006237095.1 | 11 | 1033 | |||
| XM_006237034.2 | XP_006237096.1 | 11 | 1033 | |||
| XM_008763239.1 | XP_008761461.1 | 11 | 1033 | |||
| GenBank mRNA | BC082059.1 | - | 10 | 1006 | 7132421 | |

Atg7
Genomic Map