siRNA Details
| Gene Symbol: Tsen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014057.1 | NP_001014079.1 | 2 | 158 | ||
| XM_008763240.1 | XP_008761462.1 | 2 | 181 | |||
| XM_008763241.1 | XP_008761463.1 | 2 | 220 | |||
| XM_008763242.1 | XP_008761464.1 | 2 | 182 | |||
| GenBank mRNA | BC087631.1 | - | 2 | 158 | 7319546 | |

Tsen2
Genomic Map