siRNA Details
| Gene Symbol: Ift122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057925.4 | XP_001057925.2 | 15 | 1896 | ||
| XM_006237067.1 | XP_006237129.1 | 16 | 2073 | |||
| GenBank mRNA | BC087667.1 | - | 15 | 1746 | 7317105 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift122| Gene Symbol: Ift122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057925.4 | XP_001057925.2 | 15 | 1896 | ||
| XM_006237067.1 | XP_006237129.1 | 16 | 2073 | |||
| GenBank mRNA | BC087667.1 | - | 15 | 1746 | 7317105 | |