siRNA Details
| Gene Symbol: Plxnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107881.1 | NP_001101351.1 | 5, 6 | 1683 | ||
| XM_006237035.2 | XP_006237097.1 | 5, 6 | 1951 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxnd1| Gene Symbol: Plxnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107881.1 | NP_001101351.1 | 5, 6 | 1683 | ||
| XM_006237035.2 | XP_006237097.1 | 5, 6 | 1951 | |||