siRNA Details
| Gene Symbol: Ankrd26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237217.2 | XP_006237279.1 | 7 | 1031 | ||
| XM_008763271.1 | XP_008761493.1 | 7 | 1031 | |||
| XM_232319.7 | XP_232319.5 | 7 | 1031 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd26| Gene Symbol: Ankrd26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237217.2 | XP_006237279.1 | 7 | 1031 | ||
| XM_008763271.1 | XP_008761493.1 | 7 | 1031 | |||
| XM_232319.7 | XP_232319.5 | 7 | 1031 | |||