siRNA Details
Gene Symbol: Cacna2d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191751.1 | NP_001178680.1 | 18, 19 | 1925 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacna2d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191751.1 | NP_001178680.1 | 18, 19 | 1925 |