siRNA Details
| Gene Symbol: Adipor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037979.1 | NP_001033068.1 | 1 | 70 | ||
| XM_006237183.1 | XP_006237245.1 | 2 | 376 | |||
| XM_006237184.2 | XP_006237246.1 | 2 | 247 | |||
| XM_006237185.2 | XP_006237247.1 | 2 | 262 | |||
| XM_006237186.2 | XP_006237248.1 | 2 | 391 | |||
| GenBank mRNA | AY724514.1 | - | 2 | 330 | ||
| DQ148392.1 | - | 1 | 70 | |||
| FQ232508.1 | - | 2 | 346 | |||

Adipor2
Genomic Map