siRNA Details
| Gene Symbol: Il17ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107883.2 | NP_001101353.2 | 13 | 1754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17ra| Gene Symbol: Il17ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107883.2 | NP_001101353.2 | 13 | 1754 | ||