siRNA Details
| Gene Symbol: Bcl2l13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107885.1 | NP_001101355.1 | 5 | 598 | ||
| XM_006237248.1 | XP_006237310.1 | 5 | 602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl2l13| Gene Symbol: Bcl2l13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107885.1 | NP_001101355.1 | 5 | 598 | ||
| XM_006237248.1 | XP_006237310.1 | 5 | 602 | |||