siRNA Details
| Gene Symbol: C1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134555.1 | NP_001128027.1 | 11 | 2140 | ||
| GenBank mRNA | BC098803.1 | - | 4 | 1033 | ||
| BC168673.1 | - | 11 | 2140 | 7937381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1r| Gene Symbol: C1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134555.1 | NP_001128027.1 | 11 | 2140 | ||
| GenBank mRNA | BC098803.1 | - | 4 | 1033 | ||
| BC168673.1 | - | 11 | 2140 | 7937381 | ||