siRNA Details
| Gene Symbol: Cops7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047098.2 | NP_001040563.2 | 4, 5 | 477 | ||
| NM_001164091.1 | NP_001157563.1 | 4, 5 | 493 | |||
| XM_006237367.1 | XP_006237429.1 | 4, 5 | 501 | |||
| XM_006237368.2 | XP_006237430.1 | 4, 5 | 687 | |||
| GenBank mRNA | AB259153.1 | - | 3, 4 | 205 | ||
| FM059742.1 | - | 4, 5 | 477 | |||
| FM122308.1 | - | 4, 5 | 493 | |||
| FQ215513.1 | - | 4, 5 | 501 | |||

Cops7a
Genomic Map