siRNA Details
| Gene Symbol: Dyrk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763308.1 | XP_008761530.1 | 15, 16 | 1735 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dyrk4| Gene Symbol: Dyrk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763308.1 | XP_008761530.1 | 15, 16 | 1735 | ||