siRNA Details
| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037353.1 | NP_001032430.1 | 7 | 1182 | ||
| GenBank mRNA | BC105773.1 | - | 7 | 1182 | 7930490 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etv6| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037353.1 | NP_001032430.1 | 7 | 1182 | ||
| GenBank mRNA | BC105773.1 | - | 7 | 1182 | 7930490 | |