siRNA Details
| Gene Symbol: Gprc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079890.1 | NP_001073359.1 | 2 | 989 | ||
| GenBank mRNA | BC086372.1 | - | 1 | 364 | ||
| BC128767.1 | - | 2 | 989 | 7124284 | ||
More Information
Set Membership: |
|

Gprc5a
Genomic Map