siRNA Details
| Gene Symbol: GAS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130831.1 | NP_001124303.1 | 8 | 729 | ||
| NM_003644.2 | NP_003635.2 | 4 | 478 | |||
| NM_201432.1 | NP_958836.1 | 8 | 886 | |||
| NM_201433.1 | NP_958839.1 | 8 | 916 | |||
| XM_005256831.3 | XP_005256888.1 | 9 | 827 | |||
| XM_005256832.3 | XP_005256889.1 | 8 | 697 | |||
| XM_005256833.3 | XP_005256890.1 | 7 | 804 | |||
| XM_005256834.3 | XP_005256891.1 | 7 | 630 | |||
| XM_005256835.3 | XP_005256892.1 | 7 | 574 | |||
| XM_011524044.2 | XP_011522346.1 | 8 | 721 | |||
| XM_017025239.1 | XP_016880728.1 | 8 | 750 | |||
| XM_017025240.1 | XP_016880729.1 | 7 | 533 | |||
| XM_017025241.1 | XP_016880730.1 | 7 | 509 | |||
| XM_017025242.1 | XP_016880731.1 | 7 | 683 | |||
| GenBank mRNA | AB007854.1 | - | 8 | 684 | ||
| AB209030.1 | - | 7 | 573 | |||
| AJ224876.1 | - | 4 | 501 | |||
| AK057761.1 | - | 5 | 548 | |||
| AK225165.1 | - | 8 | 708 | |||
| AK292987.1 | - | 8 | 702 | |||
| AK293755.1 | - | 4 | 309 | |||
| AK294829.1 | - | 7 | 677 | |||
| AK307987.1 | - | 9 | 738 | |||
| AK309722.1 | - | 8 | 737 | |||
| AK315162.1 | - | 4 | 491 | |||
| AY327406.1 | - | 8 | 916 | |||
| BC001152.1 | - | 8 | 709 | 3353809 | ||
| BC006454.1 | - | 8 | 886 | 3547000 | ||
| BT006891.1 | - | 7 | 563 | |||

GAS7
Genomic Map