siRNA Details
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237583.2 | XP_006237645.1 | 5 | 1159 | ||
| XM_008763452.1 | XP_008761674.1 | 5 | 443 | |||
| XM_232499.8 | XP_232499.6 | 4 | 742 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eps8| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237583.2 | XP_006237645.1 | 5 | 1159 | ||
| XM_008763452.1 | XP_008761674.1 | 5 | 443 | |||
| XM_232499.8 | XP_232499.6 | 4 | 742 | |||