siRNA Details
Gene Symbol: Recql | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012098.1 | NP_001012098.1 | 8 | 1110 | ||
XM_006237617.2 | XP_006237679.1 | 8 | 1081 | |||
XM_006237618.1 | XP_006237680.1 | 8 | 1085 | |||
XM_006237619.2 | XP_006237681.1 | 8 | 1023 | |||
XM_006237620.2 | XP_006237682.1 | 8 | 1089 | |||
XM_006237621.2 | XP_006237683.1 | 8 | 1081 | |||
XM_006237622.2 | XP_006237684.1 | 8 | 1081 | |||
XM_008763380.1 | XP_008761602.1 | 7 | 954 | |||
GenBank mRNA | BC079026.1 | - | 8 | 1110 | 7114009 |
