siRNA Details
| Gene Symbol: Cmas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009419.1 | NP_001009419.1 | 7, 8 | 1122 | ||
| GenBank mRNA | BC087599.1 | - | 7, 8 | 1122 | 7313414 | |
| FQ230721.1 | - | 6, 7 | 739 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmas| Gene Symbol: Cmas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009419.1 | NP_001009419.1 | 7, 8 | 1122 | ||
| GenBank mRNA | BC087599.1 | - | 7, 8 | 1122 | 7313414 | |
| FQ230721.1 | - | 6, 7 | 739 | |||