siRNA Details
| Gene Symbol: Tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007701.1 | NP_001007702.1 | 942 | |||
| GenBank mRNA | AY387055.1 | - | 825 | |||
| BC083851.1 | - | 942 | 7123274 | |||
| FQ223598.1 | - | 814 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tram1| Gene Symbol: Tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007701.1 | NP_001007702.1 | 942 | |||
| GenBank mRNA | AY387055.1 | - | 825 | |||
| BC083851.1 | - | 942 | 7123274 | |||
| FQ223598.1 | - | 814 | ||||