siRNA Details
| Gene Symbol: Pcmtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257345.1 | NP_001244274.1 | 5 | 881 | ||
| XM_008763511.1 | XP_008761733.1 | 7 | 2081 | |||
| XM_008763512.1 | XP_008761734.1 | 6 | 1971 | |||
| XM_008763513.1 | XP_008761735.1 | 7 | 2036 | |||
| XM_008763514.1 | XP_008761736.1 | 6 | 1926 | |||
| XM_008763515.1 | XP_008761737.1 | 5 | 2037 | |||

Pcmtd1
Genomic Map