siRNA Details
| Gene Symbol: Impad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008772.2 | NP_001008772.2 | 5 | 1051 | ||
| GenBank mRNA | AY321340.1 | - | 1248 | |||
| Gene Symbol: NEWGENE_1306455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763564.1 | XP_008761786.1 | 5 | 1095 | ||

Impad1
Genomic Map