siRNA Details
Gene Symbol: Tmem222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113780.1 | NP_001107252.1 | 5 | 479 | ||
GenBank mRNA | BC158875.1 | - | 5 | 479 | 7934037 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113780.1 | NP_001107252.1 | 5 | 479 | ||
GenBank mRNA | BC158875.1 | - | 5 | 479 | 7934037 |