siRNA Details
| Gene Symbol: Map3k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107909.1 | NP_001101379.1 | 27 | 3970 | ||
| XM_006239019.2 | XP_006239081.1 | 27 | 4736 | |||
| XM_006239020.2 | XP_006239082.1 | 26 | 4712 | |||
| XM_006239021.2 | XP_006239083.1 | 27 | 3489 | |||
| XM_006239022.2 | XP_006239084.1 | 25 | 4123 | |||
| XM_008764137.1 | XP_008762359.1 | 26 | 4709 | |||

Map3k6
Genomic Map