siRNA Details
| Gene Symbol: Eya3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107910.1 | NP_001101380.1 | 9 | 741 | ||
| XM_006239024.2 | XP_006239086.1 | 9 | 897 | |||
| XM_006239025.2 | XP_006239087.1 | 10 | 985 | |||
| XM_006239026.2 | XP_006239088.1 | 9 | 854 | |||
| XM_006239027.2 | XP_006239089.1 | 8 | 757 | |||
| XM_006239028.2 | XP_006239090.1 | 8 | 769 | |||

Eya3
Genomic Map