siRNA Details
Gene Symbol: Eya3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107910.1 | NP_001101380.1 | 5, 6 | 330 | ||
XM_006239024.2 | XP_006239086.1 | 4, 5 | 348 | |||
XM_006239025.2 | XP_006239087.1 | 5, 6 | 436 | |||
XM_006239026.2 | XP_006239088.1 | 4, 5 | 305 | |||
XM_006239027.2 | XP_006239089.1 | 4, 5 | 346 | |||
XM_006239028.2 | XP_006239090.1 | 3, 4 | 220 |
