siRNA Details
| Gene Symbol: Dnajc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013168.1 | NP_001013186.1 | 6 | 465 | ||
| GenBank mRNA | BC081863.1 | - | 6 | 465 | 7191298 | |
| FQ214236.1 | - | 6 | 467 | |||
| FQ231836.1 | - | 6 | 462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc8| Gene Symbol: Dnajc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013168.1 | NP_001013186.1 | 6 | 465 | ||
| GenBank mRNA | BC081863.1 | - | 6 | 465 | 7191298 | |
| FQ214236.1 | - | 6 | 467 | |||
| FQ231836.1 | - | 6 | 462 | |||