siRNA Details
| Gene Symbol: Phc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013169.2 | NP_001013187.2 | 13 | 2385 | ||
| XM_006238926.2 | XP_006238988.1 | 14 | 2456 | |||
| XM_006238930.2 | XP_006238992.1 | 6 | 1103 | |||
| XM_008764139.1 | XP_008762361.1 | 6 | 1068 | |||
| GenBank mRNA | BC090339.1 | - | 3 | 424 | ||
| BC169102.1 | - | 6 | 1047 | |||

Phc2
Genomic Map