siRNA Details
| Gene Symbol: Cpne3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107917.2 | NP_001101387.2 | 2 | 265 | ||
| XM_006237932.1 | XP_006237994.1 | 3 | 387 | |||
| GenBank mRNA | AM746508.1 | - | 2 | 265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpne3| Gene Symbol: Cpne3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107917.2 | NP_001101387.2 | 2 | 265 | ||
| XM_006237932.1 | XP_006237994.1 | 3 | 387 | |||
| GenBank mRNA | AM746508.1 | - | 2 | 265 | ||