siRNA Details
| Gene Symbol: CST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003650.3 | NP_003641.3 | 2 | 445 | ||
| GenBank mRNA | AB015225.1 | - | 2 | 317 | ||
| AF031824.1 | - | 2 | 384 | |||
| AF036342.1 | - | 2 | 226 | |||
| BC015507.1 | - | 2 | 340 | 3893020 | ||
| BG547285.1 | - | 2 | 446 | |||
| BT009825.1 | - | 2 | 235 | |||
| CR541860.1 | - | 2 | 169 | |||
| CR541878.1 | - | 2 | 169 | |||

CST7
Genomic Map