siRNA Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107918.1 | NP_001101388.1 | 4, 5 | 444 | ||
| XM_006237938.2 | XP_006238000.1 | 4, 5 | 472 | |||
| XM_006237939.2 | XP_006238001.1 | 4, 5 | 474 | |||
| XM_006237940.2 | XP_006238002.1 | 4, 5 | 469 | |||
| XM_006237941.2 | XP_006238003.1 | 4, 5 | 472 | |||

Usp45
Genomic Map