siRNA Details
| Gene Symbol: Fbxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107919.1 | NP_001101389.1 | 8 | 1740 | ||
| XM_006237942.1 | XP_006238004.1 | 8 | 1747 | |||
| XM_006237943.2 | XP_006238005.1 | 5 | 1430 | |||
| GenBank mRNA | BC161859.1 | - | 8 | 1747 | 7096047 | |

Fbxl4
Genomic Map