siRNA Details
| Gene Symbol: CST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003650.3 | NP_003641.3 | 3 | 605 | ||
| GenBank mRNA | AB015225.1 | - | 3 | 477 | ||
| AF031824.1 | - | 3 | 544 | |||
| AF036342.1 | - | 3 | 386 | |||
| BC015507.1 | - | 3 | 500 | 3893020 | ||
| BG547285.1 | - | 3 | 606 | |||
| BT009825.1 | - | 3 | 395 | |||
| CR541860.1 | - | 3 | 329 | |||
| CR541878.1 | - | 3 | 329 | |||

CST7
Genomic Map