siRNA Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_232840.8 | XP_232840.4 | 7 | 1274 | ||
| GenBank mRNA | BC087711.1 | - | 8 | 1101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl32| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_232840.8 | XP_232840.4 | 7 | 1274 | ||
| GenBank mRNA | BC087711.1 | - | 8 | 1101 | ||